Assay Details
Target Gene Details
Entrez Gene ID: | 20652 |
Gene Name: | sterol O-acyltransferase 1 |
Gene Aliases: |
8430426K15Rik, ACAT-1, AW550831, Acact, ald, hid |
Location: |
Chr.1:156428108-156474328 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Soat1 | NM_009230.3 | NP_033256.2 | ||
AK052299.1 | BAC34925.1 | |||
AK142486.1 | BAE25081.1 | |||
AK159393.1 | BAE35047.1 | |||
AK159529.1 | BAE35158.1 | |||
AK160017.1 | BAE35563.1 | |||
BC083092.1 | AAH83092.1 | |||
DQ903181.1 | ||||
S81092.1 | AAB36050.1 |
More Information
Set Membership: |
Intragenic Non-exonic |