Assay Details
Target Gene Details
Entrez Gene ID: | 240726 |
Gene Name: | solute carrier organic anion transporter family, member 5A1 |
Gene Aliases: |
4732454B15, A630033C23Rik, AA407151 |
Location: |
Chr.1:12866549-12992932 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slco5a1 | NM_172841.2 | 5 | 2207 | NP_766429.2 |
AK041736.1 | 5 | 2208 | BAC31048.1 | |
BC117943.1 | ||||
BC117944.1 | 3 | 1609 |
More Information
Set Membership: |
Intragenic Exonic |