Assay Details
Target Gene Details
Entrez Gene ID: | 20339 |
Gene Name: | selectin, endothelial cell |
Gene Aliases: |
CD62E, E-selectin, Elam |
Location: |
Chr.1:164039604-164058487 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sele | NM_011345.2 | NP_035475.1 | ||
AK153623.1 | BAE32123.1 | |||
BC119083.1 | ||||
BC120795.1 | ||||
M87862.1 | AAA37577.1 |