Assay Details
Target Gene Details
Entrez Gene ID: | 19264 |
Gene Name: | protein tyrosine phosphatase, receptor type, C |
Gene Aliases: |
B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc |
Location: |
Chr.1:138062859-138175755 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprc | NM_001111316.2 | 24 | 2637 | NP_001104786.2 |
NM_001268286.1 | 21 | 2246 | NP_001255215.1 | |
NM_011210.4 | 22 | 2318 | NP_035340.3 | |
AK054056.1 | 22 | 2319 | BAC35638.2 | |
AK154893.1 | 21 | 2241 | BAE32907.1 | |
M14342.1 | 22 | 2280 | AAA39458.1 | |
M14343.1 | 18 | 2811 | ||
M92933.1 | 24 | 2637 | AAA39459.1 |
More Information
Set Membership: |
Intragenic Exonic |