Assay Details
Target Gene Details
Entrez Gene ID: | 19264 |
Gene Name: | protein tyrosine phosphatase, receptor type, C |
Gene Aliases: |
B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc |
Location: |
Chr.1:138062859-138175755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprc | NM_001111316.2 | NP_001104786.2 | ||
NM_001268286.1 | NP_001255215.1 | |||
NM_011210.4 | NP_035340.3 | |||
AK054056.1 | BAC35638.2 | |||
AK154893.1 | BAE32907.1 | |||
M14342.1 | AAA39458.1 | |||
M14343.1 | ||||
M92933.1 | AAA39459.1 |
More Information
Set Membership: |
Intragenic Non-exonic |