Assay Details
Target Gene Details
Entrez Gene ID: | 18627 |
Gene Name: | period circadian clock 2 |
Gene Aliases: |
mKIAA0347, mPer2 |
Location: |
Chr.1:91415979-91459393 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Per2 | NM_011066.3 | 20 | 3321 | NP_035196.2 |
AF035830.1 | 20 | 3268 | AAC53592.1 | |
AF036893.1 | 20 | 3291 | AAC39942.1 | |
AK122253.1 | 20 | 3286 | BAC65535.1 | |
AK159847.1 | 20 | 3310 | BAE35425.1 | |
AK165556.1 | 20 | 3324 | BAE38253.1 |
More Information
Set Membership: |
Intragenic Exonic |