Assay Details
Target Gene Details
Entrez Gene ID: | 12355 |
Gene Name: | nuclear receptor subfamily 1, group I, member 3 |
Gene Aliases: |
AA209988, AI551208, CAR, CAR-beta, Care2, ESTM32, MB67 |
Location: |
Chr.1:171213935-171218845 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nr1i3 | NM_001243062.1 | NP_001229991.1 | ||
NM_001243063.1 | NP_001229992.1 | |||
NM_009803.5 | NP_033933.2 | |||
AF009327.1 | AAC53349.1 | |||
AF009328.1 | AAC53350.1 | |||
AI132241.1 | ||||
AK133515.1 | BAE21697.1 | |||
AK149381.1 | 6 | 894 | ||
BC037645.1 | AAH37645.1 | |||
BC089541.1 | AAH89541.1 |