Assay Details
Target Gene Details
Entrez Gene ID: | 227157 |
Gene Name: | membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4) |
Gene Aliases: |
Dlg6 |
Location: |
Chr.1:59120935-59163389 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpp4 | NM_001164682.1 | NP_001158154.1 | ||
NM_145143.3 | NP_660125.2 | |||
AB059357.1 | BAB69494.1 | |||
AB059358.1 | BAB69495.1 | |||
AJ748820.1 | CAG38657.1 | |||
AK044682.1 | BAC32031.1 | |||
BC061694.1 | ||||
BC116723.1 |
More Information
Set Membership: |
Intragenic Non-exonic |