Assay Details
Target Gene Details
Entrez Gene ID: | 65102 |
Gene Name: | Ngg1 interacting factor 3-like 1 (S. pombe) |
Gene Aliases: |
1110030G24Rik |
Location: |
Chr.1:58445156-58462276 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nif3l1 | NM_022988.2 | 3 | 686 | NP_075364.2 |
AF284439.1 | 4 | 1163 | AAG45961.1 | |
AK003978.1 | 4 | 794 | BAB23102.1 | |
AK011670.1 | 4 | 703 | BAB27769.1 | |
AK131631.1 | 4 | 1049 | BAE20729.1 | |
AK154815.1 | 4 | 778 | BAE32848.1 | |
AK158616.1 | 4 | 1069 | BAE34583.1 | |
BC065163.1 | 3 | 686 | AAH65163.1 | |
BC089475.1 | 4 | 613 | AAH89475.1 |