Assay Details
Target Gene Details
Entrez Gene ID: | 16782 |
Gene Name: | laminin, gamma 2 |
Gene Aliases: |
AA589349 |
Location: |
Chr.1:153122756-153186447 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamc2 | NM_008485.3 | 7 | 1012 | NP_032511.3 |
L20477.1 | 5 | 572 | AAA68093.1 | |
U43327.2 | 6 | 745 | AAA85256.2 |
More Information
Set Membership: |
Intragenic Exonic |