Assay Details
Target Gene Details
Entrez Gene ID: | 69312 |
Gene Name: | protein phosphatase 1, regulatory subunit 42 |
Gene Aliases: |
1700011J18Rik, 4930418G15Rik, Lrrc67, TLRR |
Location: |
Chr.1:9968622-10013541 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r42 | NM_145692.2 | 6 | 780 | NP_663730.1 |
NR_110971.1 | 6 | 795 | ||
AK015164.1 | 6 | 790 | BAB29732.1 | |
AK076637.1 | 6 | 781 | BAC36427.1 | |
BC022722.1 | 6 | 764 | AAH22722.1 |
More Information
Set Membership: |
Intragenic Exonic |