Assay Details
Target Gene Details
Entrez Gene ID: | 67444 |
Gene Name: | integrin-linked kinase-associated serine/threonine phosphatase 2C |
Gene Aliases: |
0710007A14Rik, 1600009O09Rik, PP2C-DELTA |
Location: |
Chr.1:91373846-91398841 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilkap | NM_023343.2 | NP_075832.1 | ||
AF117831.1 | AAD17234.1 | |||
AK003007.1 | ||||
AK005408.1 | ||||
BC026953.1 | AAH26953.1 | |||
BC027439.1 |
More Information
Set Membership: |
Intragenic Non-exonic |