Assay Details
Target Gene Details
Entrez Gene ID: | 74106 |
Gene Name: | DDB1 and CUL4 associated factor 6 |
Gene Aliases: |
1200006M05Rik, Iqwd1, PC326 |
Location: |
Chr.1:165329500-165460553 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcaf6 | NM_028759.1 | 6 | 860 | NP_083035.1 |
AK004618.1 | 6 | 723 | BAB23414.1 | |
AK213381.1 | 2 | 94 |
More Information
Set Membership: |
Intragenic Exonic |