Assay Details
Target Gene Details
Entrez Gene ID: | 15926 |
Gene Name: | isocitrate dehydrogenase 1 (NADP+), soluble |
Gene Aliases: |
AI314845, AI788952, E030024J03Rik, Id-1, Idh-1, Idpc |
Location: |
Chr.1:65158616-65186479 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Idh1 | NM_001111320.1 | 5 | 795 | NP_001104790.1 |
NM_010497.3 | 5 | 773 | NP_034627.3 | |
AF020039.1 | 4 | 623 | AAD02919.1 | |
AK087063.1 | 5 | 675 | BAC39792.1 | |
AK149019.1 | 5 | 704 | BAE28720.1 | |
AK151212.1 | 5 | 698 | BAE30207.1 | |
AK159173.1 | 5 | 704 | BAE34873.1 | |
AK160896.1 | 5 | 705 | BAE36075.1 | |
AK167158.1 | 5 | 745 | BAE39299.1 | |
AK167583.1 | 5 | 709 | BAE39644.1 | |
AK168832.1 | 5 | 747 | ||
BC088986.1 | 5 | 745 | AAH88986.1 | |
CF950262.1 | ||||
DQ403075.1 | 4 | 523 |
More Information
Set Membership: |
Intragenic Exonic |