Assay Details
Target Gene Details
Entrez Gene ID: | 208727 |
Gene Name: | histone deacetylase 4 |
Gene Aliases: |
4932408F19Rik, AI047285 |
Location: |
Chr.1:91928779-92180385 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac4 | NM_207225.1 | NP_997108.1 | ||
AK029933.1 | BAE43272.1 | |||
AK155250.1 | BAE33147.1 | |||
BC066052.1 | AAH66052.1 |
More Information
Set Membership: |
Intragenic Non-exonic |