Assay Details
Target Gene Details
Entrez Gene ID: | 227095 |
Gene Name: | 3-hydroxyisobutyryl-Coenzyme A hydrolase |
Gene Aliases: |
2610509I15Rik, AI648812 |
Location: |
Chr.1:52845046-52920860 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hibch | NM_146108.1 | NP_666220.1 | ||
AK076038.1 | BAC36138.1 | |||
AK082965.1 | ||||
AK186066.1 | ||||
AK206848.1 | ||||
AK214768.1 | ||||
BC026437.1 | AAH26437.1 |
More Information
Set Membership: |
Intragenic Non-exonic |