Assay Details
Target Gene Details
Entrez Gene ID: | 226861 |
Gene Name: | hedgehog acyltransferase |
Gene Aliases: |
2810432O22Rik, AI462858, AP-2CRE, Skn, Tg(TFAP2A-cre)1Will |
Location: |
Chr.1:192512828-192771412 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hhat | NM_144881.4 | NP_659130.2 | ||
AK028230.1 | BAC25829.1 | |||
AK052669.1 | BAC35090.1 | |||
AK134661.1 | ||||
AK172126.1 | BAE42837.1 |
More Information
Set Membership: |
Intragenic Non-exonic |