Assay Details
Target Gene Details
Entrez Gene ID: | 226866 |
Gene Name: | somatomedin B and thrombospondin, type 1 domain containing |
Gene Aliases: |
Gm106, Rpesp |
Location: |
Chr.1:15853862-15892722 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sbspon | NM_001033288.3 | 4 | 708 | NP_001028460.1 |
AK137144.1 | 4 | 709 | BAE23250.1 | |
AK143251.1 | 4 | 682 | BAE25326.1 | |
BC147194.1 | 4 | 695 | ||
BC147195.1 | 4 | 695 |