Assay Details
Target Gene Details
Entrez Gene ID: | 67698 |
Gene Name: | family with sequence similarity 174, member A |
Gene Aliases: |
2310044D20Rik, Fam174, Naa6, Tmem157 |
Location: |
Chr.1:95313628-95335284 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam174a | NM_026321.4 | NP_080597.2 | ||
AB154249.1 | ||||
AK009797.1 | BAB26509.1 | |||
AK009892.1 | BAB26567.1 | |||
AK017318.1 | BAB30690.1 | |||
AK020633.1 | BAB32159.1 | |||
AK033948.1 | BAC28523.1 | |||
AK084861.1 | 2 | 722 | ||
AK148807.1 | BAE28669.1 | |||
BC021428.1 | AAH21428.1 |