Assay Details
Target Gene Details
Entrez Gene ID: | 269109 |
Gene Name: | dipeptidylpeptidase 10 |
Gene Aliases: |
6430601K09Rik, DPP X, Dprp3 |
Location: |
Chr.1:123332138-124845960 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp10 | NM_199021.3 | 20 | 1925 | NP_950186.3 |
AK032579.1 | 18 | 1571 | ||
AK046842.1 | 20 | 1926 | ||
BC029696.1 | 5 | 407 | AAH29696.1 | |
BC063074.1 | 10 | 847 | AAH63074.1 | |
BC067026.1 | 20 | 1918 | AAH67026.1 |
More Information
Set Membership: |
Intragenic Exonic |