Assay Details
Target Gene Details
Entrez Gene ID: | 227325 |
Gene Name: | delta/notch-like EGF-related receptor |
Gene Aliases: |
A930026D19Rik, BET, Bret |
Location: |
Chr.1:84369839-84696221 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dner | NM_152915.1 | 6 | 1566 | NP_690879.1 |
AB067650.1 | 6 | 1215 | BAB72175.1 | |
AF370126.1 | 6 | 1222 | AAM14419.1 | |
AK044597.1 | 6 | 1329 | BAC31995.1 | |
AK152755.1 | BAE31470.1 | |||
AK153235.1 | BAE31828.1 | |||
AY032924.1 | 6 | 1222 | AAK50342.1 | |
BC022636.1 | AAH22636.1 | |||
BC034634.1 | 6 | 1566 | AAH34634.1 |