Assay Details
Target Gene Details
Entrez Gene ID: | 227325 |
Gene Name: | delta/notch-like EGF-related receptor |
Gene Aliases: |
A930026D19Rik, BET, Bret |
Location: |
Chr.1:84369839-84696221 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dner | NM_152915.1 | 5 | 1416 | NP_690879.1 |
AB067650.1 | 5 | 1065 | BAB72175.1 | |
AF370126.1 | 5 | 1072 | AAM14419.1 | |
AK030642.1 | 5 | 1215 | ||
AK044597.1 | 5 | 1179 | BAC31995.1 | |
AY032924.1 | 5 | 1072 | AAK50342.1 | |
BC034634.1 | 5 | 1416 | AAH34634.1 |