Assay Details
Target Gene Details
Entrez Gene ID: | 13211 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 9 |
Gene Aliases: |
AI326842, Ddx9, HEL-5, NDHII, RHA, mHEL-5 |
Location: |
Chr.1:153455756-153487686 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx9 | NM_007842.2 | 15 | 1758 | NP_031868.2 |
AK135603.1 | 10 | 1073 | ||
AK141824.1 | 15 | 1753 | BAE24846.1 | |
AK148423.1 | 15 | 1754 | ||
AY512925.1 | 11 | 1214 | AAR87796.1 | |
BC089159.1 | 15 | 1747 | AAH89159.1 | |
U91922.1 | 15 | 1715 | AAC05725.1 | |
U92080.1 | 4 | 453 | AAB72087.1 |
More Information
Set Membership: |
Intragenic Exonic |