Assay Details
Target Gene Details
Entrez Gene ID: | 12283 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
AA408805, AA960512, C78372, MO25, MO25alpha |
Location: |
Chr.1:85793410-85851577 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cab39 | NM_133781.4 | NP_598542.3 | ||
AK144665.1 | BAE25997.1 | |||
BC020041.1 | AAH20041.1 | |||
S51858.1 | AAB24801.1 |