Assay Details
Target Gene Details
Entrez Gene ID: | 12269 |
Gene Name: | complement component 4 binding protein |
Gene Aliases: |
AI195242, C4bpa |
Location: |
Chr.1:130635921-130661618 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C4bp | NM_007576.3 | 2 | 260 | NP_031602.3 |
AB094486.1 | 2 | 176 | BAC75872.1 | |
AK136735.1 | 2 | 174 | BAE23113.1 | |
AK149468.1 | 2 | 267 | BAE28899.1 | |
AK149477.1 | 2 | 177 | BAE28904.1 | |
AK162345.1 | 2 | 174 | ||
BC012257.1 | 2 | 232 | AAH12257.1 | |
M17122.1 | 2 | 274 | AAA37312.1 |