Assay Details
Target Gene Details
Entrez Gene ID: | 51800 |
Gene Name: | BCL2-related ovarian killer |
Gene Aliases: |
AI847676, AU021146, matador, mtd |
Location: |
Chr.1:93685694-93695762 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bok | NM_016778.2 | NP_058058.1 | ||
AF027707.1 | AAC53582.1 | |||
AK156243.1 | BAE33639.1 | |||
AK168373.1 | BAE40305.1 | |||
AK171258.1 | BAE42349.1 | |||
BC030069.1 | AAH30069.1 |