Assay Details
Target Gene Details
Entrez Gene ID: | 108664 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit H |
Gene Aliases: |
0710001F19Rik, AU022349, CGI-11, SFD, SFDalpha, SFDbeta, VMA13 |
Location: |
Chr.1:5082978-5162549 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v1h | NM_133826.4 | NP_598587.2 | ||
AK034165.1 | ||||
AK078767.1 | BAC37382.1 | |||
AK081492.1 | ||||
AK135137.1 | ||||
AK151659.1 | BAE30587.1 | |||
AK152147.1 | ||||
BC009154.1 | AAH09154.1 |
More Information
Set Membership: |
Intragenic Non-exonic |