Assay Details
Target Gene Details
Entrez Gene ID: | 72949 |
Gene Name: | cyclin T2 |
Gene Aliases: |
2900041I18Rik, C81304, CycT2, CycT2a, CycT2b |
Location: |
Chr.1:127774164-127804837 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnt2 | NM_028399.1 | 7 | 643 | NP_082675.1 |
AK047125.1 | 3 | 1543 | ||
AK049863.1 | 7 | 588 | ||
AK152399.1 | 7 | 591 | BAE31187.1 | |
AK154439.1 | 7 | 601 | BAE32588.1 | |
BC054122.1 | 7 | 643 | AAH54122.1 | |
JX965957.1 | 7 | 579 | ||
JX965958.1 | 7 | 579 |
More Information
Set Membership: |
Intragenic Exonic |