Assay Details
Target Gene Details
Entrez Gene ID: | 11363 |
Gene Name: | acyl-Coenzyme A dehydrogenase, long-chain |
Gene Aliases: |
AA960361, AU018452, C79855, LCAD |
Location: |
Chr.1:66830839-66863309 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acadl | NM_007381.4 | NP_031407.2 | ||
AK005140.1 | BAB23838.1 | |||
AK018319.1 | BAB31161.1 | |||
AK151845.1 | ||||
AK167537.1 | ||||
BC027412.1 | AAH27412.1 | |||
U21489.1 | AAC52329.1 |
More Information
Set Membership: |
Intragenic Non-exonic |