Assay Details
Target Gene Details
Entrez Gene ID: | 226409 |
Gene Name: | zinc finger, RAN-binding domain containing 3 |
Gene Aliases: |
4933425L19Rik, AH2, AI316834, C730006D09 |
Location: |
Chr.1:127954179-128103049 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zranb3 | NM_001285945.1 | NP_001272874.1 | ||
NM_027678.3 | NP_081954.1 | |||
AK016921.1 | BAB30499.1 | |||
AK042139.1 | BAC31181.1 | |||
AK050039.1 | BAC34042.1 | |||
AK083641.1 | BAC38978.1 | |||
AK084943.1 | BAC39315.1 | |||
AK085488.1 | ||||
BC066035.1 | AAH66035.1 | |||
BC117921.1 | ||||
BC117922.2 |
More Information
Set Membership: |
Intragenic Non-exonic |