Assay Details
Target Gene Details
Entrez Gene ID: | 227059 |
Gene Name: | solute carrier family 39 (zinc transporter), member 10 |
Gene Aliases: |
2900042E17Rik, 5430433I10, mKIAA1265 |
Location: |
Chr.1:46807544-46925772 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc39a10 | NM_172653.2 | NP_766241.2 | ||
AK049099.1 | BAC33542.1 | |||
AK122483.1 | BAC65765.1 | |||
AK136117.1 | 7 | 1880 | ||
BC052880.1 | AAH52880.1 | |||
BC059214.1 | AAH59214.1 | |||
BC062918.1 | AAH62918.1 |
More Information
Set Membership: |
Intragenic Non-exonic |