Assay Details
Target Gene Details
Entrez Gene ID: | 226407 |
Gene Name: | RAB3 GTPase activating protein subunit 1 |
Gene Aliases: |
1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130 |
Location: |
Chr.1:127868737-127943876 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab3gap1 | NM_178690.4 | 9 | 833 | NP_848805.2 |
AK076244.1 | 9 | 797 | BAC36271.1 | |
AK076399.1 | 9 | 816 | BAC36323.1 | |
AK080432.1 | 9 | 815 | BAC37915.1 | |
AK134604.1 | 9 | 815 | BAE22204.1 | |
AK164523.1 | 9 | 814 | BAE37821.1 | |
AK170768.1 | 9 | 805 | ||
AK172881.1 | 9 | 820 | BAD32159.1 | |
BC046297.1 | 9 | 799 | AAH46297.1 |
More Information
Set Membership: |
Intragenic Exonic |