Assay Details
Target Gene Details
Entrez Gene ID: | 68421 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
0910001K20Rik, AV347960 |
Location: |
Chr.1:24678544-24766301 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmbrd1 | NM_026719.2 | NP_080995.2 | ||
AK152170.1 | BAE31002.1 | |||
AK153237.1 | BAE31829.1 | |||
AK158982.1 | BAE34756.1 | |||
AK167080.1 | BAE39240.1 | |||
AY061989.1 | AAL38052.1 | |||
BC025579.1 | AAH25579.1 | |||
BC027774.1 | AAH27774.1 | |||
BC031902.1 | AAH31902.1 | |||
BC039053.1 | AAH39053.1 |
More Information
Set Membership: |
Intragenic Non-exonic |