Assay Details
Target Gene Details
Entrez Gene ID: | 16782 |
Gene Name: | laminin, gamma 2 |
Gene Aliases: |
AA589349 |
Location: |
Chr.1:153122756-153186447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamc2 | NM_008485.3 | NP_032511.3 | ||
AF106279.1 | 1 | 129 | AAF14184.1 | |
DV654715.1 | ||||
DV658346.1 | ||||
U43327.2 | AAA85256.2 |
More Information
Set Membership: |
Intragenic Non-exonic |