Assay Details
Target Gene Details
Entrez Gene ID: | 14125 |
Gene Name: | Fc receptor, IgE, high affinity I, alpha polypeptide |
Gene Aliases: |
FcERI, Fce1a, Fcr-5, fcepsilonri |
Location: |
Chr.1:173221269-173227232 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcer1a | NM_010184.2 | NP_034314.2 | ||
BC125455.1 | ||||
BC125457.1 | ||||
CJ142154.1 | ||||
CJ142191.1 | ||||
J05018.1 | AAA37600.1 |
More Information
Set Membership: |
Intragenic Non-exonic |