Assay Details
Target Gene Details
Entrez Gene ID: | 211383 |
Gene Name: | APC membrane recruitment 3 |
Gene Aliases: |
9430069J07Rik, Fam123c |
Location: |
Chr.1:34578935-34590948 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amer3 | NM_213727.2 | NP_998892.1 | ||
BC070435.1 | AAH70435.1 | |||
BY290690.1 |