Assay Details
Target Gene Details
Entrez Gene ID: | 103967 |
Gene Name: | dynamin 3 |
Gene Aliases: |
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820 |
Location: |
Chr.1:161982450-162478323 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnm3 | NM_001038619.1 | NP_001033708.1 | ||
NM_172646.2 | NP_766234.1 | |||
AK036199.1 | BAC29343.1 | |||
AK049724.1 | BAC33895.1 | |||
AK082698.1 | BAC38575.1 | |||
AK122379.1 | BAC65661.1 | |||
AK147678.1 | BAE28068.1 | |||
BC141143.1 | ||||
BC141144.1 |
More Information
Set Membership: |
Intragenic Non-exonic |