Assay Details
Target Gene Details
Entrez Gene ID: | 100502745 |
Gene Name: | RIKEN cDNA 2900009J06 gene |
Gene Aliases: |
- |
Location: |
Chr.1:127753617-127774054 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
2900009J06Rik | NR_045298.1 | 2 | 198 | |
AK013506.1 | 2 | 202 | ||
BB663189.1 | 2 | 220 | ||
BX636135.1 | 1 | 271 |
Target Gene Details
Entrez Gene ID: | 266645 |
Gene Name: | amino carboxymuconate semialdehyde decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.1:127729413-127767564 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acmsd | NM_001033041.2 | NP_001028213.1 | ||
AB071419.1 | BAB86939.1 | |||
AK143962.1 | BAE25634.1 | |||
BC139281.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |