Assay Details
Target Gene Details
Entrez Gene ID: | 12028 |
Gene Name: | BCL2-associated X protein |
Gene Aliases: |
- |
Location: |
Chr.7:45461695-45466903 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bax | NM_007527.3 | 3 | 251 | NP_031553.1 |
AK013298.1 | 3 | 161 | BAB28776.1 | |
AK149994.1 | 3 | 144 | BAE29222.1 | |
AK159233.1 | 3 | 216 | BAE34919.1 | |
AY095934.1 | 2 | 598 | AAM34436.1 | |
BC018228.1 | 3 | 196 | AAH18228.1 | |
BC053380.1 | 3 | 168 | AAH53380.1 | |
BY308683.1 | 3 | 231 | ||
CJ078904.1 | 3 | 252 | ||
L22472.1 | 3 | 128 | AAA03622.1 |
More Information
Additional Information:
Set Membership: |
Intragenic Exonic Validated |