Assay Details
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcor | NM_001168321.1 | 4 | 2967 | NP_001161793.1 |
NM_029510.3 | 4 | 3636 | NP_083786.2 | |
NM_175044.3 | 4 | 3636 | NP_778209.2 | |
NM_175045.3 | 4 | 3636 | NP_778210.2 | |
NM_175046.3 | 4 | 3636 | NP_778211.2 | |
AK053309.1 | BAC35338.1 | |||
AK086385.2 | 4 | 3143 | BAE43390.1 | |
AK129398.1 | 4 | 2967 | BAC98208.1 | |
AY161170.1 | 4 | 2951 | AAN85318.1 | |
AY161171.1 | 4 | 2951 | AAN85319.1 | |
AY161172.1 | 4 | 2951 | AAN85320.1 | |
AY161173.1 | 4 | 2951 | AAN85321.1 | |
BC058656.1 | 4 | 3137 | AAH58656.1 |