Assay Details
Target Gene Details
Entrez Gene ID: | 70495 |
Gene Name: | ATPase, H+ transporting, lysosomal accessory protein 2 |
Gene Aliases: |
(P)RR, 5730403E06Rik, APT6M8-9, ATP6M8-9, Atp6ip2, M8-9 |
Location: |
Chr.X:12587759-12617051 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6ap2 | NM_027439.4 | NP_081715.1 | ||
AB192471.2 | ||||
AK017482.1 | BAB30766.2 | |||
AK029405.1 | BAC26436.1 | |||
AK076495.1 | BAC36365.1 | |||
AK151735.1 | ||||
AK168139.1 | ||||
BC014706.1 | AAH14706.1 | |||
BY784908.1 |
More Information
Set Membership: |
Intragenic Non-exonic |