Assay Details
Target Gene Details
Entrez Gene ID: | 71458 |
Gene Name: | BCL6 interacting corepressor |
Gene Aliases: |
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575 |
Location: |
Chr.X:12036737-12160355 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcor | NM_001168321.1 | 11 | 4604 | NP_001161793.1 |
NM_029510.3 | 12 | 5429 | NP_083786.2 | |
NM_175044.3 | 11 | 5375 | NP_778209.2 | |
NM_175045.3 | 12 | 5327 | NP_778210.2 | |
NM_175046.3 | 11 | 5273 | NP_778211.2 | |
AK018034.1 | 2 | 195 | ||
AK018370.1 | 4 | 684 | ||
AK031282.1 | 4 | 731 | ||
AK129398.1 | 11 | 4604 | BAC98208.1 | |
AY161170.1 | 12 | 4744 | AAN85318.1 | |
AY161171.1 | 11 | 4690 | AAN85319.1 | |
AY161172.1 | 12 | 4642 | AAN85320.1 | |
AY161173.1 | 11 | 4588 | AAN85321.1 | |
BC058656.1 | 12 | 4828 | AAH58656.1 |