Assay Details
Target Gene Details
Entrez Gene ID: | 60364 |
Gene Name: | downstream neighbor of SON |
Gene Aliases: |
1110025J21Rik, AI845729, ORF60 |
Location: |
Chr.16:91666856-91688759 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Donson |
Target Gene Details
Entrez Gene ID: | 20658 |
Gene Name: | Son DNA binding protein |
Gene Aliases: |
2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp |
Location: |
Chr.16:91647824-91679195 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Son | NM_178880.4 | 9 | 7063 | NP_849211.3 |
AK034390.1 | 1 | 2759 | ||
AK035150.1 | 2 | 2554 | BAC28960.1 | |
AK122418.1 | 8 | 4179 | BAC65700.1 | |
CB849818.2 | 2 | 161 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |