Assay Details
Target Gene Details
Entrez Gene ID: | 20658 |
Gene Name: | Son DNA binding protein |
Gene Aliases: |
2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp |
Location: |
Chr.16:91647824-91679195 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Son | NM_019973.2 | 4 | 6390 | NP_064357.2 |
NM_178880.4 | 4 | 6390 | NP_849211.3 | |
AB546195.1 | 5 | 6205 | ||
AF193607.1 | 5 | 6214 | AAF23121.1 | |
AK040621.1 | 2 | 1393 | BAC30648.1 | |
AK122418.1 | 2 | 751 | BAC65700.1 |
More Information
Set Membership: |
Intragenic Exonic |