Assay Details
Target Gene Details
Entrez Gene ID: | 22129 |
Gene Name: | tetratricopeptide repeat domain 3 |
Gene Aliases: |
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119 |
Location: |
Chr.16:94370739-94469222 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc3 | NM_009441.2 | 2 | 162 | NP_033467.2 |
AB008516.1 | 1 | 39 | BAA31563.1 | |
AK053765.1 | 2 | 266 | BAC35513.1 | |
AK084201.1 | ||||
AK088273.1 | 2 | 163 | BAC40250.1 | |
AK089974.1 | 2 | 105 | BAC41023.1 | |
AK165850.1 | BAE38412.1 | |||
AK166120.1 | ||||
AK166164.1 | ||||
AK166298.1 | 2 | 104 | ||
AK170087.1 | 2 | 153 | ||
AK220495.1 | 2 | 204 | BAD90293.1 | |
BC019173.1 | 2 | 131 | ||
BC049277.1 | 2 | 144 | ||
BC057207.1 | 2 | 120 | AAH57207.1 |
More Information
Set Membership: |
Intragenic Exonic |