Assay Details
Target Gene Details
Entrez Gene ID: | 22129 |
Gene Name: | tetratricopeptide repeat domain 3 |
Gene Aliases: |
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119 |
Location: |
Chr.16:94370739-94469222 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc3 | NM_009441.2 | 22 | 2072 | NP_033467.2 |
AB008516.1 | 21 | 1949 | BAA31563.1 | |
AK053765.1 | 22 | 2176 | BAC35513.1 | |
AK084201.1 | 12 | 1107 | ||
AK088273.1 | 21 | 2019 | BAC40250.1 | |
AK165850.1 | 11 | 980 | BAE38412.1 | |
AK166120.1 | 12 | 1012 | ||
AK166164.1 | 13 | 1161 | ||
AK220495.1 | 22 | 2114 | BAD90293.1 | |
BC019173.1 | 22 | 2041 | ||
BC057207.1 | 21 | 1976 | AAH57207.1 |
More Information
Set Membership: |
Intragenic Exonic |