Assay Details
Target Gene Details
Entrez Gene ID: | 93871 |
Gene Name: | bromodomain and WD repeat domain containing 1 |
Gene Aliases: |
5330419I02Rik, D530019K20Rik, G1-403-16, Wdr9, repro5 |
Location: |
Chr.16:95992089-96082428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brwd1 | NM_001103179.1 | NP_001096649.1 | ||
NM_145125.3 | NP_660107.2 | |||
AJ292467.1 | CAC44373.1 | |||
AJ292468.1 | CAC44374.1 | |||
CF162350.1 | ||||
CV558558.1 |