Assay Details
Target Gene Details
Entrez Gene ID: | 54720 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
Adapt78, CALP1L, CSP1, DSC1, Dscr1, MCIP1, RCN1 |
Location: |
Chr.16:92391951-92466169 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rcan1 | NM_001081549.2 | 4 | 797 | NP_001075018.1 |
NM_019466.4 | 4 | 643 | NP_062339.2 | |
AF237789.1 | 4 | 547 | AAF63485.1 | |
AF237790.1 | 4 | 547 | AAF63486.1 | |
AF260717.1 | 4 | 561 | AAF70343.1 | |
AF263239.1 | 4 | 577 | AAF72701.1 | |
AF263240.1 | 4 | 660 | AAF72702.1 | |
AF282255.1 | 4 | 554 | AAF91461.1 | |
AK077283.1 | 4 | 643 | BAC36729.1 | |
AK146764.1 | 4 | 772 | BAE27416.1 | |
AY325904.1 | 4 | 753 | AAP96744.1 | |
BC013551.1 | 4 | 623 | AAH13551.1 |
More Information
Set Membership: |
Intragenic Exonic |