Assay Details
Target Gene Details
Entrez Gene ID: | 14450 |
Gene Name: | phosphoribosylglycinamide formyltransferase |
Gene Aliases: |
Gaps, Prgs |
Location: |
Chr.16:91621395-91647481 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gart | NM_010256.2 | 15 | 1997 | NP_034386.2 |
AK146355.1 | 15 | 1982 | BAE27107.1 | |
AK146478.1 | 15 | 1939 | ||
AK168458.1 | 15 | 1957 | ||
AK168501.1 | 15 | 1960 | BAE40386.1 | |
AK168724.1 | 15 | 1972 | BAE40565.1 | |
AK168796.1 | 15 | 1972 | BAE40629.1 | |
AK168864.1 | 15 | 1948 | BAE40683.1 | |
AK168876.1 | 15 | 1949 | BAE40694.1 | |
AK185268.1 | 2 | 272 | ||
AK203694.1 | 2 | 218 | ||
BC070465.1 | 15 | 1944 | AAH70465.1 | |
U01024.1 | 14 | 1902 | AAA19013.1 |