Assay Details
Target Gene Details
Entrez Gene ID: | 54720 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
Adapt78, CALP1L, CSP1, DSC1, Dscr1, MCIP1, RCN1 |
Location: |
Chr.16:92391951-92466169 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rcan1 | NM_001081549.2 | 1 | 255 | NP_001075018.1 |
AF237789.1 | AAF63485.1 | |||
AF260717.1 | AAF70343.1 | |||
AF282255.1 | AAF91461.1 | |||
AK146764.1 | 1 | 230 | BAE27416.1 | |
AY325904.1 | 1 | 211 | AAP96744.1 | |
BY276032.1 | 1 | 256 |
More Information
Set Membership: |
Intragenic Exonic |